Please login to your online account to display your discounted pricing

Buffer solution, pH 10.00 (+/-0.01 at 25°C), Colored Blue, Specpure™, NIST Traceable, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Buffer solution
Name Note Colored Blue, Specpure™, NIST Traceable
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA38709AP

  SDS
alfa aesar
38709AP
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

pH 10.00 (± 0.01 at 25°C)
Quantity 500mL
Chemical Name or Material Buffer solution
Color Blue
TSCA TSCA
Name Note Colored Blue, Specpure™, NIST Traceable
Physical Form Liquid
MDL Number MFCD00146206
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse. Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse. Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.