Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Certified, LabChem™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: lab chem  LC125001

 View more versions of this product

Catalog No. LC125001

  • / Each
 

Description & Specifications

Chemical Identifiers

pH 10.00 ± 0.02
Purity Grade Certified Grade
Formula Weight 381.37
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
Health Hazard 1 Danger
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Decomposition Information Boron
Identification Passes Test
Recommended Storage Room Temperature
CAS 1303-96-4
CAS Max % 99.57
Linear Formula Na2B4O7·10H2O
CAS 7732-18-5
Quantity 500mL
Solubility Soluble in water
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Chemical Name or Material Buffer Solution pH 10.0 at 25°C
Physical Form Liquid
Color clear, colorless solution

Specifications

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
CAS Max % 99.57
CAS 1303-96-4
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature