Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Certified, LabChem™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: lab chem  LC125101

 View more versions of this product

Catalog No. LC125101

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
Name Note Color Coded Blue
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 99.57
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
CAS 1303-96-4
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
CAS Max % 0.38
Recommended Storage Room Temperature
CAS 1310-73-2
CAS Max % 0.05
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Decomposition Information Boron
pH 10.00 ± 0.02
Color clear, blue solution
Quantity 500mL

Specifications

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
CAS Max % 99.57
CAS 1303-96-4
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Name Note Color Coded Blue
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature