missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Certified, LabChem™

Manufacturer:  Lab Chem LC125105

 View more versions of this product

Catalog No. LC125105


 

Specifications Description & Specifications

Specifications

Buffer Solution pH 10.0 at 25°C
7732-18-5,1303-96-4,1310-73-2
Certified Grade
Na2B4O7·10H2O
Passes Test
Soluble in water
Boron
clear, blue solution
Color Coded Blue
99.57,0.38,0.05
Liquid
381.37
Cubitainer™
Traceable to NIST
10.00 ± 0.02
20L

Chemical Identifiers

7732-18-5,1303-96-4,1310-73-2
Na2B4O7·10H2O
Buffer Solution pH 10.0 at 25°C, Color Coded Blue
99.57,0.38,0.05
381.37
Certified Grade

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

Danger

Recommended Storage : Room Temperature