Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Certified, LabChem™


Quantity 1L
Packaging Poly Bottle

Manufacturer: lab chem  LC125102

 View more versions of this product

Catalog No. LC125102

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 381.37
pH 10.00 ± 0.02
Purity Grade Certified Grade
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
Health Hazard 1 Danger
Color clear, blue solution
CAS Max % 0.05
CAS 1303-96-4
Packaging Poly Bottle
CAS Max % 0.38
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
Chemical Name or Material Buffer Solution pH 10.0 at 25°C
Decomposition Information Boron
Name Note Color Coded Blue
CAS Max % 99.57
Identification Passes Test
Traceability to NIST Traceable to NIST
Quantity 1L
Recommended Storage Room Temperature
CAS 7732-18-5
CAS 1310-73-2
Solubility Soluble in water

Specifications

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
CAS Max % 99.57
CAS 1303-96-4
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Name Note Color Coded Blue
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature