Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Certified, LabChem™

$15.74 - $118.69

Chemical Identifiers

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
CAS Max % 99.57
CAS 1303-96-4
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Name Note Color Coded Blue
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC125101

 
lab chem
LC125101
Poly Bottle 500mL Each for $15.74

LC125102

 
lab chem
LC125102
Poly Bottle 1L Each for $22.57

LC1251020

 
lab chem
LC1251020
Pack 20 x 20mL Case of 20 Each for $94.40

LC125104

 
lab chem
LC125104
Poly Bottle 4L Each for $37.49

LC125104C

 
lab chem
LC125104C
Cubitainer™ 4L Each for $71.86

LC125105

 
lab chem
LC125105
Cubitainer™ 20L Each for $118.69

LC125106

 
lab chem
LC125106
Cubitainer™ 10L Each for $76.69
Description & Specifications

Specifications

pH 10.00 ± 0.02
Purity Grade Certified Grade
Formula Weight 381.37
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
Chemical Name or Material Buffer Solution pH 10.0 at 25°C
Decomposition Information Boron
CAS Max % 0.05
Color clear, blue solution
Quantity 500mL
Name Note Color Coded Blue
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
CAS 7732-18-5
Identification Passes Test
CAS Max % 99.57
CAS 1303-96-4
Packaging Poly Bottle
CAS Max % 0.38
Recommended Storage Room Temperature
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS 1310-73-2
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature