Please login to your online account to display your discounted pricing

Buffer Solution pH 10.0 at 25°C, Certified, LabChem™

$14.60 - $89.86

Chemical Identifiers

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
CAS Max % 99.57
CAS 1303-96-4
CAS Max % 0.38
CAS 1310-73-2
CAS Max % 0.05
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC125001

 
lab chem
LC125001
Poly Bottle 500mL Each for $14.60

LC125002

 
lab chem
LC125002
Poly Bottle 1L Each for $22.51

LC125004

 
lab chem
LC125004
Poly Bottle 4L Each for $31.60

LC125005

 
lab chem
LC125005
Cubitainer™ 20L Each for $89.86

LC125006

 
lab chem
LC125006
Cubitainer™ 10L Each for $63.97
Description & Specifications

Specifications

Chemical Name or Material Buffer Solution pH 10.0 at 25°C
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 99.57
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
CAS 1303-96-4
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
CAS Max % 0.38
Recommended Storage Room Temperature
CAS 1310-73-2
CAS Max % 0.05
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Decomposition Information Boron
pH 10.00 ± 0.02
Color clear, colorless solution
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature