missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 1.68 at 25°C, Certified, LabChem™
Supplier: LabChem LC122104
Specifications
Buffer Solution pH 1.68 at 25°C | |
98.55,1.26,0.19 | |
Potassium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors | |
1.68 ±0.01 | |
4 L | |
HO2CCO2H | |
1g/mL | |
Soluble in water | |
Certified |
Colorless | |
6153-56-6,1310-58-3 | |
1g/mL | |
Liquid | |
Passes Test | |
Poly Bottle | |
Traceable to NIST | |
90.04 |