Please login to your online account to display your discounted pricing

Buffer Solution pH 1.68, Alfa Aesar™ Specpure™

$32.26 - $88.28

Chemical Identifiers

Chemical Name or Material Buffer solution
CAS 7447-40-7
CAS 7647-01-0
CAS 7732-18-5
Physical Form Liquid
MDL Number MFCD00147253
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42415AP

  SDS
alfa aesar
42415AP
500mL Each for $32.26

AA42415K7

  SDS
alfa aesar
42415K7
4L Each for $88.28

AA42415K2

  SDS
alfa aesar
42415K2
1L Each for $48.53
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Density 1.005g/cm3 at 20°C
Chemical Name or Material Buffer solution
MDL Number MFCD00147253
CAS 7447-40-7
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%
Appearance Colorless liquid
Health Hazard 1 H318-H315: Causes serious eye damage. Causes skin irritation.
CAS 7647-01-0
Odor Odorless
CAS 7732-18-5
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Safety and Handling

H318-H315: Causes serious eye damage. Causes skin irritation.