Please login to your online account to display your discounted pricing

Buffer solution, pH 1.00 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

For Calibration of pH Meters

Chemical Identifiers

Chemical Name or Material Buffer solution
CAS 7447-40-7
CAS 7647-01-0
CAS 7732-18-5
Physical Form Liquid
MDL Number MFCD00146252
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40446AP

  SDS
alfa aesar
40446AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40446K2

  SDS
alfa aesar
40446K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40446K7

  SDS
alfa aesar
40446K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Buffer solution
MDL Number MFCD00146252
CAS 7447-40-7
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%
Appearance Colorless liquid
Health Hazard 1 H318-H315: Causes serious eye damage. Causes skin irritation.
CAS 7647-01-0
CAS 7732-18-5
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Safety and Handling

H318-H315: Causes serious eye damage. Causes skin irritation.