missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution pH 1.0 at 25°C, Certified, LabChem™

Manufacturer:  LabChem LC121804

 View more versions of this product

Catalog No. LC121804



Specifications

Specifications

Buffer Solution pH 1.0 at 25°C
7447-40-7,7647-01-0
UN1789
74.55
Certified
Soluble in water
1g/mL
Chlorine; Thermal decomposition generates: Corrosive vapors
Colorless
99.58,0.37,0.05
KCl
Passes Test
Liquid
Poly Bottle
Traceable to NIST
1g/mL
1.00 ±0.02
4L
Documents
Promotions

Promotions