missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical

$131.39 - $131.39

Chemical Identifiers

CAS 7732-18-5,7784-31-8,10294-26-5,5329-14-6,10043-35-3,7732-18-5 (Water, Deionized)
CAS Min % 96.0,1.66,0.33,0.24,0.12
CAS Max % 100.0,1.73,0.34,0.25,0.13
Physical Form Liquid
Chemical Name or Material Buffer Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
1473-16 Ricca Chemical Company
RI147316
Glass amber 500mL Each for $131.39
1473-32 Ricca Chemical Company
RI147332
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Nitrate Determination using Ion Selective Electrode
Description & Specifications

Chemical Identifiers

7732-18-5,7784-31-8,10294-26-5,5329-14-6,10043-35-3,7732-18-5 (Water, Deionized)
100.0,1.73,0.34,0.25,0.13
Buffer Solution
96.0,1.66,0.33,0.24,0.12
Liquid

Specifications

7732-18-5,7784-31-8,10294-26-5,5329-14-6,10043-35-3,7732-18-5 (Water, Deionized)
100.0,1.73,0.34,0.25,0.13
Glass amber
500mL
96.0,1.66,0.33,0.24,0.12
Liquid
Clear colorless
Buffer Solution