Please login to your online account to display your discounted pricing

Buffer Solution, Giordano, pH 6.4, for Wright Staining, Ricca Chemical


Quantity 4L
Packaging Poly natural

Manufacturer: ricca chemical company  RI14501

 View more versions of this product

Catalog No. RC14501

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Buffer Solution, Giordano
CAS 7732-18-5
CAS 7558-79-4
CAS 7778-77-0
CAS Min % 97.0
CAS Min % 0.25
CAS Max % 0.26
CAS Max % 100.0
CAS Min % 0.64
CAS Max % 0.67
Packaging Poly natural
Name Note pH 6.4

Specifications

Chemical Name or Material Buffer Solution, Giordano
Name Note pH 6.4
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7778-77-0
CAS Min % 0.64
CAS Max % 0.67
CAS 7558-79-4
CAS Min % 0.25
CAS Max % 0.26
Physical Form Liquid