missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, Giordano, pH 6.4, for Wright Staining, Ricca Chemical

$32.91 - $114.09

Chemical Identifiers

CAS 7732-18-5,7778-77-0,7558-79-4
CAS Min % 97.0,0.64,0.25
CAS Max % 100.0,0.67,0.26
Physical Form Liquid
Chemical Name or Material Buffer Solution, Giordano, pH 6.4
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RC14501 Ricca Chemical Company
RI14501
Poly natural 4L Each for $114.09
RC145016 Ricca Chemical Company
RI145016
Poly natural 500mL Each for $32.91
Specifications

Description

Wright Staining
Description & Specifications

Chemical Identifiers

7732-18-5,7778-77-0,7558-79-4
100.0,0.67,0.26
Buffer Solution, Giordano, pH 6.4
97.0,0.64,0.25
Liquid

Specifications

7732-18-5,7778-77-0,7558-79-4
100.0,0.67,0.26
Poly natural
4L
97.0,0.64,0.25
Liquid
Clear colorless
Buffer Solution, Giordano, pH 6.4