missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, Giordano, pH 6.4, for Wright's Stain, Certified, LabChem™

Manufacturer:  LabChem LC125901

 View more versions of this product

Catalog No. LC125901



Specifications

Specifications

Buffer Solution, Giordano
pH 6.4, for Wright's Stain
7778-77-0,7558-79-4,50-00-0,67-56-1
Passes Test
KH2PO4
1g/mL
136.09
Certified
6.40 to 6.45
500mL
98.975,0.66,0.32,0.04,0.005
1g/mL
Poly Bottle
Soluble in water
Carbon monoxide; Carbon dioxide
Liquid
Colorless
Documents
Promotions

Promotions