missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution (Borate), pH 10, Color Coded Blue, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  Avantor Performance 565501

 View more versions of this product

Catalog No. 02002481



Specifications

Specifications

Buffer Solution (Borate), pH 10
BAKER ANALYZED™ Reagent
1L = 1.00kg
Color Coded Blue
Poly Bottle
500mL
Documents