Please login to your online account to display your discounted pricing

Buffer Solution (Borate), pH 10, Color Coded Blue, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$69.55 - $739.02

Chemical Identifiers

Chemical Name or Material Buffer Solution (Borate), pH 10
Name Note Color Coded Blue
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002481

 
avantor performance
565501
Poly Bottle 500mL Each for $69.55

02002484

 
avantor performance
565503
Cubetainer 4L Each for $739.02
Description & Specifications

Specifications

Chemical Name or Material Buffer Solution (Borate), pH 10
Name Note Color Coded Blue
Purity Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Quantity 500mL