Please login to your online account to display your discounted pricing

Buffer Solution B, for Sulfate, Certified, LabChem™

$139.86 - $139.86

Chemical Identifiers

Chemical Name or Material Buffer Solution B, for Sulfate
CAS 7732-18-5
CAS Max % 94.39
CAS 7791-18-6
CAS Max % 3
CAS 64-19-7
CAS Max % 2
CAS 6131-90-4
CAS Max % 0.5
CAS 7757-79-1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC125752

 
lab chem
LC125752
1L Each for $139.86
Description & Specifications

Specifications

Purity Grade Certified Grade
pH 3.5 ± 0.5
Formula Weight 203.31
CAS 6131-90-4
Solubility Soluble in water
Identification Passes Test
CAS Max % 0.5
CAS 7732-18-5
CAS Max % 3
CAS 7757-79-1
CAS Max % 0.1
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
Health Hazard 1 Warning
CAS Max % 94.39
Linear Formula MgCl2·6H2O
Physical Form Liquid
Packaging Poly Bottle
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Color Clear, colorless
CAS 64-19-7
Quantity 1L
CAS 7791-18-6
Chemical Name or Material Buffer Solution B, for Sulfate
CAS Max % 2
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Traceability to NIST Traceable to NIST
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation and serious eye irritation.

Room Temperature