missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution B, for low level Sulfate Analysis (Turbidimetric Method), Ricca Chemical

For Analysis of Low-Level Sulfate (Turbidimetric Method)

Chemical Identifiers

CAS 7732-18-5,7791-18-6,64-19-7,6131-90-4,7757-79-1,7757-82-6
CAS Min % 93.0,2.78,1.95,0.46,0.09,0.01
CAS Max % 96.0,2.9,2.03,0.48,0.1,0.01
Physical Form Liquid
Chemical Name or Material Buffer Solution B
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
1476-32 Ricca Chemical Company
RI147632
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14768 Ricca Chemical Company
RI14768
Poly natural 250mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7791-18-6,64-19-7,6131-90-4,7757-79-1,7757-82-6
96.0,2.9,2.03,0.48,0.1,0.01
Buffer Solution B
93.0,2.78,1.95,0.46,0.09,0.01
Liquid

Specifications

Liquid
1L
7732-18-5,6131-90-4,7757-82-6,64-19-7,7791-18-6,7757-79-1
Poly natural
Clear colorless
96.0,2.9,0.48,0.1,0.01,2.03
0.46,2.78,1.95,0.01,0.09,93.0
Buffer Solution B