missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution A, for Sulfate, Certified, LabChem™

$142.89 - $142.89

Chemical Identifiers

CAS 7732-18-5,7791-18-6,64-19-7,6131-90-4,7757-79-1
CAS Max % 94.4,3,2,0.5,0.1
Linear Formula MgCl2·6H2O
Formula Weight 203.31
Chemical Name or Material Buffer Solution A, for Sulfate
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC125702 Lab Chem
LC125702
1L Each for $142.89
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7791-18-6,64-19-7,6131-90-4,7757-79-1
MgCl2·6H2O
Buffer Solution A, for Sulfate
94.4,3,2,0.5,0.1
203.31
Certified Grade

Specifications

7732-18-5,7791-18-6,64-19-7,6131-90-4,7757-79-1
Liquid
203.31
Poly Bottle
Traceable to NIST
3.5 ± 0.5
1L
Certified Grade
94.4,3,2,0.5,0.1
MgCl2·6H2O
Passes Test
Soluble in water
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Clear, colorless
Buffer Solution A, for Sulfate

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature