missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 9.18 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI15955

 View more versions of this product

Catalog No. R15955



Specifications

Specifications

Buffer, Reference Standard
1303-96-4,7732-18-5
100.0,0.39
Cubitainer
20L
pH 9.18± 0.01 at 25°C
98.0,0.37
Liquid
Colorless
Documents
Promotions

Promotions