missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 9.18 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 159532
Specifications
Buffer, Reference Standard | |
1303-96-4,7732-18-5 | |
Colorless | |
9.18 | |
1 L | |
Non-sterile |
pH 9.18± 0.01 at 25°C | |
98.0,0.37 | |
100.0,0.39 | |
Liquid | |
Natural Poly Bottle | |
Reference Standard |