missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 159032
Specifications
Buffer, Reference Standard | |
10043-35-3,7447-40-7,1310-73-2,7732-18-5 | |
Liquid | |
Natural Poly Bottle |
pH 9.00± 0.01 at 25°C | |
Colorless | |
1 L |