missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 8.00 ± 0.01 at 25°C, Ricca Chemical

$66.47 - $330.52

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 8.00± 0.01 at 25°C
CAS 7778-77-0,7558-79-4,7732-18-5
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1580-1
View Documents
Ricca Chemical Company
RI15801
4L Poly Bottle
Each for $160.88
 
1580-25
View Documents
Ricca Chemical Company
RI158025
10L Cubitainer™
Each for $242.61
 
36-101-8159
View Documents
Ricca Chemical Company
RI158032
1L Poly Bottle
Each for $66.47
 
1580-5
View Documents
Ricca Chemical Company
RI15805
20L Cubitainer™
Each for $330.52
 
Description

Description

Specifications

Specifications

Buffer, Reference Standard
pH 8.00± 0.01 at 25°C
7778-77-0,7558-79-4,7732-18-5
SDS