missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 7.70 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI15761

 View more versions of this product

Catalog No. 15761



Specifications

Specifications

Buffer, Reference Standard
7558-79-4,7778-77-0,7732-18-5
100.0,0.9,0.17
Poly natural
4L
pH 7.70± 0.01 at 25 °C
97.0,0.87,0.16
Liquid
Colorless
Documents
Promotions

Promotions