missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 7.40 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI15651

 View more versions of this product

Catalog No. 15651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Buffer, Reference Standard
7558-79-4,7778-77-0,7732-18-5
100.0,0.9,0.29
Poly natural
4L
pH 7.40± 0.01 at 25°C
97.0,0.86,0.28
Liquid
Colorless
Documents