missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow), Ricca Chemical

Manufacturer:  Ricca Chemical Company RI155120

 View more versions of this product

Catalog No. 155120



Specifications

Specifications

Buffer, Reference Standard
7558-79-4,7778-77-0,7732-18-5
100.0,0.91,0.5
20mL
pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow)
97.0,0.87,0.48
Pouch (20/Box)
Documents