missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), Ricca Chemical
Supplier: Ricca Chemical Company 15511CT
Specifications
Buffer, Reference Standard | |
7558-79-4,7778-77-0,7732-18-5 | |
100.0,0.91,0.5 | |
Cubitainer |
pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow) | |
97.0,0.87,0.48 | |
4 L |