missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 7.00 + 0.01 at 25°C (Color Coded Green), Ricca Chemical

Manufacturer:  Ricca Chemical Company RI155145

 View more versions of this product

Catalog No. 15514-5



Specifications

Specifications

Buffer, Reference Standard
97.0,0.87,0.48
Liquid
Green
7558-79-4,7778-77-0,7732-18-5
100.0,0.9,0.5
Cubitainer™
20L
Documents