missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 6.86 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company 15405

 View more versions of this product

Catalog No. 1540-5



Specifications

Specifications

Buffer, Reference Standard
7558-79-4,7778-77-0,7732-18-5
100.0,0.36,0.34
Cubitainer
20L
pH 6.86± 0.01 at 25°C
97.0,0.34,0.33
Liquid
Colorless
Documents