missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 6.00 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI151025

 View more versions of this product

Catalog No. 1510-25



Specifications

Specifications

Buffer, Reference Standard
7778-77-0,7558-79-4,7732-18-5
100.0,0.82,0.11
Cubitainer™
10L
pH 6.00± 0.01 at 25°C
97.0,0.78,0.1
Liquid
Colorless
Documents