missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 5.00 0.01 at 25°C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI15051

 View more versions of this product

Catalog No. 1505-1



Specifications

Specifications

Buffer, Reference Standard
877-24-7,7732-18-5,7732-18-5
Poly Natural
4L
pH 5.00 ± 0.01 at 25°C
Liquid
Colorless
Documents