missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 5.00 0.01 at 25°C, Ricca Chemical

$37.32 - $198.73

Chemical Identifiers

CAS 1310-73-2 (Sodium Hydroxide),877-24-7 (Potassium Acid Phthalate),7732-18-5 (Water, Deionized)
Physical Form Liquid
Chemical Name or Material Buffer, Reference Standard, pH 5.00 ± 0.01 at 25°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
1505-1 Ricca Chemical Company
RI15051
Poly Natural 4L Each for $135.03
1505-25 Ricca Chemical Company
RI15052.5
Cubitainer™ 10L Each for $198.73
36-101-8138 Ricca Chemical Company
RI150516
Poly Natural 500mL Each for $37.32
36-101-8139 Ricca Chemical Company
RI150532
Poly Natural 1L Each for $44.26
Specifications Description & Specifications

Chemical Identifiers

1310-73-2 (Sodium Hydroxide),877-24-7 (Potassium Acid Phthalate),7732-18-5 (Water, Deionized)
Liquid
Buffer, Reference Standard, pH 5.00 ± 0.01 at 25°C

Specifications

1310-73-2 (Sodium Hydroxide),877-24-7 (Potassium Acid Phthalate),7732-18-5 (Water, Deionized)
Poly Natural
4L
Liquid
Clear colorless
Buffer, Reference Standard, pH 5.00 ± 0.01 at 25°C