missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 5.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 150532
Specifications
Buffer, Reference Standard | |
877-24-7,7732-18-5 | |
Liquid | |
Natural Poly Bottle |
pH 5.00 ± 0.01 at 25°C | |
Colorless | |
1 L |