missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 4.00 (Color Coded Red), Ricca Chemical

Manufacturer:  Ricca Chemical Company RI150120

 View more versions of this product

Catalog No. 150120



Specifications

Specifications

Buffer, Reference Standard
877-24-7,7732-18-5
100.0,1.03
20 x 20mL
pH 4.00 ± 0.01 at 25 deg C (Color Coded Red)
97.0,0.99
Pouch
Documents