missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C (Color Coded Red), Ricca Chemical
$56.84 - $1165.41
Specifications
Chemical Name or Material | Buffer, Reference Standard |
---|---|
Name Note | pH 4.00 ± 0.01 at 25 deg C (Color Coded Red) |
CAS | 877-24-7,7732-18-5 |
CAS Min % | 97.0,0.99 |
CAS Max % | 100.0,1.03 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
150120
|
Ricca Chemical Company
150120 |
20 x 20 mL | Pack |
Each for $189.50
|
|
||||
1501-16
|
Ricca Chemical Company
R1501000500A |
500 mL | Natural Poly Bottle |
Each for $56.84
|
|
||||
150132
|
Ricca Chemical Company
150132 |
1 L | Natural Poly Bottle |
Each for $78.72
|
|
||||
15011CT
|
Ricca Chemical Company
15011CT |
4 L | Cubitainer |
Each for $236.41
|
|
||||
1501-1
|
Ricca Chemical Company
15011 |
4 L | Natural Poly Bottle |
Each for $193.56
|
|
||||
15-012-00
|
Ricca Chemical Company
150120B |
200 x 20 mL | Pack |
Each for $1,165.41
|
|
||||
1501-25
|
Ricca Chemical Company
150125 |
10 L | Cubitainer |
Each for $189.81
|
|
||||
1501-5
|
Ricca Chemical Company
R150100020F |
20 L | Cubitainer |
Each for $289.47
|
|
||||
Specifications
Buffer, Reference Standard | |
877-24-7,7732-18-5 | |
100.0,1.03 |
pH 4.00 ± 0.01 at 25 deg C (Color Coded Red) | |
97.0,0.99 |