missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer, Reference Standard, pH 4.00 ± 0.01 at 25 deg C, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI15005

 View more versions of this product

Catalog No. 1500-5



Specifications

Specifications

Buffer, Reference Standard
877-24-7,7732-18-5
100.0,1.03
20L
pH 4.00 ± 0.01 at 25 deg C
97.0,0.99
Cubitainer™
Documents