missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 150025
Specifications
Buffer, Reference Standard | |
877-24-7,7732-18-5 | |
100.0,1.03 | |
Cubitainer |
pH 4.00 ± 0.01 at 25 deg C | |
97.0,0.99 | |
10 L |