missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical
Supplier: Ricca Chemical Company 160125
Specifications
Buffer, Reference Standard | |
0°C | |
100°C | |
Blue | |
10 | |
10 L | |
Miscible |
pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) | |
497-19-8,144-55-8,7732-18-5 | |
97.0,0.51,0.39 | |
100.0,0.53,0.41 | |
Liquid | |
Cubitainer |