Please login to your online account to display your discounted pricing

Buffer Concentrate, pH 7.00 (Certified), Fisher Chemical


Quantity 1L
Packaging Poly Bottle

Manufacturer: fisher scientific company  BPSB109-1

 View more versions of this product

Catalog No. SB109-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 1L
NIST Traceable to NIST.
Physical Form Liquid
CAS 7664-38-2
Identification Pass Test
CAS 7732-18-5
Packaging Poly Bottle
CAS 1310-73-2
MDL Number MFCD00146206
CAS 1310-58-3
Chemical Name or Material Buffer Concentrate, pH 7.00
CAS 64-02-8
ChemAlert Storage Symbol Gray
Traceability to NIST Pass Test

Specifications

Chemical Name or Material Buffer Concentrate, pH 7.00
CAS 64-02-8
CAS 7732-18-5
CAS 1310-58-3
CAS 1310-73-2
CAS 7664-38-2
Purity Grade Certified Grade
Physical Form Liquid
MDL Number MFCD00146206
Accessories
 

SDS & Safety Documents