Please login to your online account to display your discounted pricing

Buffer Concentrate, pH 4.00 (Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  BPSB99-500

 View more versions of this product

Catalog No. SB99-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Buffer Concentrate, pH 4.00
Additional Information Vapor Pressure: 14mmHg at 20°C
CAS 877-24-7
ChemAlert Storage Symbol Gray
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified Grade
Physical Form Liquid
MDL Number MFCD00013070
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST Pass Test
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Analyte or Component Names Potassium Acid Phthalate
NIST Traceable to NIST.
Boiling Point 100°C
Melting Point 0°C
pH 4.0
Color Clear
Quantity 500mL

Specifications

Chemical Name or Material Buffer Concentrate, pH 4.00
CAS 877-24-7
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified Grade
Physical Form Liquid
MDL Number MFCD00013070
Accessories
 

SDS & Safety Documents