BUCHI Receiving Flasks for Rotary Evaporators
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Manufacturer:  BUCHI 000426
 View more versions of this product
Catalog No. 05-402-656
Description
Description
  • Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Constant monitoring of glass wall thickness uniformity
  • High degree of attention paid to sphericity of rotating glass parts
  • Glass parts are tension-relieved at 560°C
  • Available in volumes from 50mL to 3000mL
  • Available uncoated or with P+G coating
Specifications
Specifications
2000mL
Rotary evaporator
Standard Glass
Ball joint KS-35
2000mL
Receiving Flask
Uncoated
Round
Documents