Please login to your online account to display your discounted pricing
BUCHI™ Receiving Flasks for Rotary Evaporators
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Manufacturer: buchi™  040776
 View more versions of this product
Catalog No. 05-402-533
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 2000mL
For Use With (Equipment) Rotary evaporator
Shape Round
Volume (English) 2000mL
Material (Coating) Plastic and Glass
Includes Ball joint KS-35
Product Type Receiving Flask