Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/V-700/V-801, Woulff bottle and w/o Glassware/Adapter


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21099C11

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-750

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators w/V-700/V-801, Woulff bottle and w/o Glassware/Adapter
Certifications CE, CSA, ISO I4000
Hertz 50/60Hz
Length (English) 5.74 in.
Length (Metric) 40cm
Voltage 220/240V
Weight (Metric) 21kg
Width (Metric) 27cm
Volume (English) 34 oz.
Weight (English) 46.3 lb.
Model P-6
Temperature Range (Metric) 95°C
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Maximum Speed Range 0 to 485rpm
Wattage 800w
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Height (English) 5.74 in.
Height (Metric) 40cm
Width (English) 10.6 in.
Height (English) Footprint 15.7 in.
Includes V-700 pump, V-801 controller, woulff bottle, 1L receiving flask
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Volume (Metric) 1000mL