Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/V-700/V-801, Woulff bottle and w/o Glassware/Adapter


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21099C21

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-753

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators w/V-700/V-801, Woulff bottle and w/o Glassware/Adapter
Certifications CE, CSA, ISO I4000
Volume (English) 67.5 oz.
Length (Metric) 40cm
Width (Metric) 27cm
Height (English) 5.74 in.
Weight (English) 46.3 lb.
Width (English) 10.6 in.
Model P-6
Length (Metric) Footprint 26cm
Capacity (Metric) 10 to 160mL
Temperature Range (Metric) 95°C
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Maximum Speed Range 0 to 485rpm
Volume (Metric) 2000mL
Height (English) Footprint 15.7 in.
Temperature Range (English) 203°F
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Wattage 800w
For Use With (Application) High throughput labs requiring multiple sample processing
Includes V-700 pump, V-801 controller, woulff bottle, 2L receiving flask
Voltage 220/240V
Hertz 50/60Hz
Height (Metric) 40cm
Weight (Metric) 21kg
Length (English) 5.74 in.