Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/V-700/V-801, Woulff bottle for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21001C11

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-737

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporators with V-700/V-801, Woulff bottle
Certifications CE, CSA, ISO I4000
Height (Metric) 40cm
Hertz 50/60Hz
Length (English) 5.74 in.
Length (Metric) 40cm
Voltage 220/240V
Width (Metric) 27cm
Width (English) 10.6 in.
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Height (Metric) Footprint 40cm
Wattage 800w
Volume (English) 0.26 gal.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Volume (Metric) 1000mL
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Model P-6
Temperature Range (Metric) 95°C
Height (English) Footprint 15.7 in.
Includes V-700 pump, V-801 controller, woulff bottle, 1L receiving flask
Length (English) Footprint 10.2 in.
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Height (English) 5.74 in.