Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/V-700/V-801, Woulff bottle for 220-240V


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21001C21

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-740

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor™ Parallel Evaporators with V-700/V-801, Woulff bottle
Certifications CE, CSA, ISO I4000
Width (Metric) 27cm
Holds 150mL Tube
Maximum Speed Range 0 to 485rpm
Height (English) Footprint 15.7 in.
Length (English) 5.74 in.
Height (Metric) 40cm
Width (English) 10.6 in.
Height (English) 5.74 in.
Length (Metric) Footprint 26cm
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Volume (English) 0.53 gal.
Height (Metric) Footprint 40cm
Volume (Metric) 2000mL
Length (English) Footprint 10.2 in.
Capacity (Metric) 10 to 160mL
Temperature Range (English) 203°F
Wattage 800w
For Use With (Application) High throughput labs requiring multiple sample processing
Includes V-700 pump, V-801 controller, woulff bottle, 2L receiving flask
Hertz 50/60Hz
Temperature Range (Metric) 95°C
Weight (English) 46.3 lb.
Length (Metric) 40cm
Voltage 220/240V
Weight (Metric) 21kg
Model P-6