Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/Safety Shield, w/o Pump, Controller, Glassware/Adapter


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21199C20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-778

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators w/Safety Shield, w/o Pump, Controller, Glassware/Adapter
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Volume (English) 67.5 oz.
Length (English) 5.74 in.
Width (Metric) 27cm
Length (Metric) 40cm
Weight (Metric) 21kg
For Use With (Application) High throughput labs requiring multiple sample processing
Model P-6
Temperature Range (Metric) 95°C
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 220-240V 50/60Hz
Includes Safety shield, 2L receiving flask
Length (English) Footprint 10.2 in.
Temperature Range (English) 203°F
Height (English) Footprint 15.7 in.
Capacity (Metric) 10 to 160mL
Height (Metric) Footprint 40cm
Volume (Metric) 2000mL
Wattage 800w
Length (Metric) Footprint 26cm
Weight (English) 46.3 lb.
Height (English) 5.74 in.
Height (Metric) 40cm
Voltage 220/240V
Hertz 50/60Hz
Width (English) 10.6 in.