Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/Safety Shield, V-700/V-801, Sec. Condenser


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP21199C22

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-780

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Parallel Evaporators w/Safety Shield, V-700/V-801, Sec. Condenser
Certifications CE, CSA, ISO I4000
Width (Metric) 27cm
Volume (English) 67.5 oz.
Hertz 50/60Hz
Maximum Speed Range 0 to 485rpm
Voltage 220/240V
Width (English) 10.6 in.
Height (English) 5.74 in.
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Weight (English) 46.3 lb.
Electrical Requirements 220-240V 50/60Hz
For Use With (Equipment) 2L receiving flask
Capacity (Metric) 10 to 160mL
Length (Metric) Footprint 26cm
Temperature Range (English) 203°F
Height (English) Footprint 15.7 in.
Includes Safety shield, V-700 pump, V-855 controller, woulff bottle, secondary condenser
Volume (Metric) 2000mL
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Wattage 800w
For Use With (Application) High throughput labs requiring multiple sample processing
Length (English) 5.74 in.
Length (Metric) 40cm
Weight (Metric) 21kg
Height (Metric) 40cm
Temperature Range (Metric) 95°C
Model P-6