Please login to your online account to display your discounted pricing

BUCHI™ Multivapor™ P-6 Parallel Evaporators w/S Condenser, w/o Adapters, Pump or Controller


Evaporates up to 6 samples on one compact unit

Manufacturer: buchi™  MP22199S20

 View more versions of this product

 Alert

Direct exposure of plastic-coated glassware to chlorinated solvents and aggressive reagents should be avoided.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 05-404-720

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Multivapor P-6 Parallel Evaporation System With S Condenser
Certifications CE, CSA, ISO I4000
Maximum Speed Range 0 to 485rpm
Volume (English) 67.5 oz.
Hertz 50/60Hz
Voltage 100/120V
Length (English) 5.74 in.
Length (Metric) 40cm
Model P-6
Includes Safety shield, 2L recieving flask
Height (Metric) Footprint 40cm
Length (English) Footprint 10.2 in.
Temperature Range (English) 203°F
Height (English) 5.74 in.
Height (Metric) 40cm
Weight (English) 46.3 lb.
Weight (Metric) 21kg
Width (Metric) 27cm
Width (English) 10.6 in.
Temperature Range (Metric) 95°C
Capacity (Metric) 10 to 160mL
For Use With (Application) High throughput labs requiring multiple sample processing
Height (English) Footprint 15.7 in.
Length (Metric) Footprint 26cm
Wattage 800w
Volume (Metric) 2000mL
Dimensions (L x W x H) 5.74 x 10.6 x 5.74 in. (40 x 27 x 40cm)
Electrical Requirements 100-120V 50/60Hz